missing translation for 'onlineSavingsMsg'
Learn More

CD3e Hamster anti-Mouse, PE-Cy7, Clone: 145-2C11, BD

Hamster Monoclonal Antibody

Marque:  BD Biosciences 561100

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15870769

  • 151.98€ / 1 pièce
Date d’expédition estimée: 15-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The 145-2C11 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex that is expressed on thymocytes, mature T lymphocytes, and NK-T cells. The cytoplasmic domain of CD3e participates in the signal transduction events that activate several cellular biochemical pathways as a result of antigen recognition. Soluble 145-2C11 antibody can activate either unprimed (naive) or primed (memory/preactivated) T cells in vivo or in vitro, in the presence of Fc receptor-bearing accessory cells. In contrast, plate-bound 145-2C11 can activate T cells in the absence of accessory cells. Soluble 145-2C11 antibody has been reported to induce re-directed lysis of Fc receptor-bearing target cells by CTL clones and can also block lysis of specific target cells by antigen-specific CTL's. Under some conditions, T-cell activation by 145-2C11 antibody has been reported to result in apoptotic cell death. The 145-2C11 antibody does not cross-react with rat leukocytes. Preincubation of thymus cell suspensions at 37°C for 2-4 hours prior to staining reportedly enhances the ability of anti-CD3ε and anti-αβ TCR mAbs to detect the T-cell receptor on immature thymocytes.

Spécification

Spécification

CD3e
145-2C11
PE-Cyanine7
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Affinity Purified
H-2Kb specific cytotoxic T lymphocyte clone BM10-37
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.