Learn More
CD41 Monoclonal Antibody (PM6/248), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA180666
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD41 antibody, clone PM6/248 recognizes the human CD41 cell surface antigen, a approximately 140 kDa glycoprotein expressed by platelets and megakaryocytes.
CD41 Monoclonal antibody specifically detects CD41 in Baboon, Human samples. It is validated for Flow Cytometry.Spécification
CD41 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
P08514 | |
Itga2b | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Baboon, Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
PM6/248 | |
FITC | |
Itga2b | |
AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
Mouse | |
Protein A | |
RUO | |
3674 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |