missing translation for 'onlineSavingsMsg'
Learn More

CD44 Rat anti-Mouse, V450, Clone: IM7, BD

Rat Monoclonal Antibody

Marque:  BD Biosciences 560451

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15870209

  • 575.00€ / 1 pièce
Expédition estimée: 23-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The IM7 antibody reacts with an epitope on both alloantigens and all isoforms of the CD44 glycoprotein (Pgp-1, Ly-24). The standard form of CD44, lacking variable exons and referred to as CD44H or CD44s, is widely expressed on hematopoietic and non-hematopoietic cells. CD44 isoforms encoded by variable exons are expressed on epithelial cells, but only at low levels on most leukocytes. Mice with the Ly-24.1 alloantigen (e.g., BALB/c, CBA/J, DBA/1, DBA/2) have relatively large subsets of CD44H+ T lymphocytes, while Ly-24.2 strains (e.g., A, AKR, CBA/N, C3H/He, C57BL, C57BR, C57L, C58, NZB, SJL, SWR, 129) have few CD44H+ T cells. CD44 is a cell adhesion receptor, and its principal ligand, hyaluronate, is a common component of extracellular matrices. Differential glycosylation of CD44 influences its binding to hyaluronate. Additional ligands include the cell-surface form of CD74 and the cytokine osteopontin (Eta-1). Bone marrow- and thymus-derived progenitor cells capable of repopulating the thymus express CD44. In the periphery, the level of CD44 expression increases upon activation of B lymphocytes, CD4+ T cells, and CD8+ T cells; memory cells can be recognized by their CD44(hi) phenotype. The IM7 mAb inhibits established collagen-induced arthritis in DBA/1 mice. Moreover, it prevents CNS inflammation and clinical symptoms of experimental autoimmune encephalomyelitis. In contrast, the same antibody exacerbates experimental autoimmune thyroiditis in CBA/J mice. The IM7 mAb recognizes a different epitope from that recognized by mAb KM114 (Mfr. No. 558739), and the antibody pair can be used in ELISA to detect soluble CD44. It has been observed that IM7 antibody cross-reacts with human, dog, cat, horse, cow, and pig leukocytes. Anti-human CD44, clone G44-26 (Mfr. No. 555476), and IM7 antibody compete for binding to human peripheral blood lymphocytes. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Rat
Clone: IM7
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M2

Flow Cytometry

Spécification

Spécification

CD44
Monoclonal
0.2mg/mL
Pgp-1; Ly-24; H-CAM; HERMES; ECMR-III; Hyaluronate Receptor
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
IM7
Violet Fluor 450 (V450)
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1
0.1 mg
Immunology
Mouse
IgG2b κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.