missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44v10 Monoclonal Antibody (VFF-14), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA516963
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD44v10 Monoclonal antibody specifically detects CD44v10 in Human samples. It is validated for Flow Cytometry, Western Blot.Spécification
CD44v10 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P16070 | |
CD44 | |
proprietary | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
VFF-14 | |
Unconjugated | |
CD44 | |
AL133330.1; AU023126; AW121933; AW146109; Cd44; CD44 antigen; CD44 molecule (Indian blood group); CD44 splice variant; CD44 var; CD44 variant; CD44A; CD44v10; CDw44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; epican; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; H-CAM v10; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCHI; HUTCH-I; Hyaluronate receptor; IN; LHR; Ly-24; Lymphocyte antigen 24; MC56; MDU2; MDU3; METAA; MGC10468; MIC4; MUTCH-I; PGP I; Pgp1; PGP-1; PGP-I; phagocytic glycoprotein 1; Phagocytic glycoprotein I; RHAMM; soluble CD44 | |
Mouse | |
purified | |
RUO | |
960 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |