missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Monoclonal Antibody (MEM-56), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110238
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry.Spécification
CD45RA | |
Monoclonal | |
PerCP | |
PTPRC | |
B220; CD45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; LCA; L-CA; leucocyte common antigen; Leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; Lyt-4; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T100; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5788 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-56 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08575 | |
PTPRC | |
Human thymocytes and T lymphocytes. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |