missing translation for 'onlineSavingsMsg'
Learn More

CD5 Mouse anti-Human, FITC, Clone: UCHT2, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 555352

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 15868088

  • 271.32€ / 1 pièce
Date d’expédition estimée: 21-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spécification

Spécification

CD5
UCHT2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
FITC
Affinity Purified
Human T cells
100 Tests
Immunology
Primary
Human
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.