missing translation for 'onlineSavingsMsg'
Learn More

CD56 Mouse anti-Human, Alexa Fluor 700, Clone: B159, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 557919

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15869108

  • 416.16€ / 1 pièce
Date d’expédition estimée: 24-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.

Spécification

Spécification

CD56
B159
Alexa Fluor 700
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39
Affinity Purified
Human NK Cells
0.1mg
Immunology
Primary
Human
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.