missing translation for 'onlineSavingsMsg'
Learn More

Cdk2 Mouse, Unlabeled, Clone: 55, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 610146

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15845289

  • 596.22€ / 1 kit
Expédition estimée: 27-02-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cyclin-dependent kinase 2 (Cdk2) is a member of a family of cdc2-related cell cycle protein kinases. Cdk2 shares 60% identity with cdc2 and its activity is regulated by phosphorylation in a similar fashion. Cdk2 is expressed earlier in the cell cycle than is cdc2. Like p34 [cdc2], p33 [cdk2] associates with Cyclin A in human cells. However, kinase activity associated with Cyclin A-Cdk2 is present in S phase, whereas, the kinase activity associated with Cyclin A-cdc2 is found only in G2. Cdk2 can also complex with cyclins E, D1, and D3. It is not known if the D cyclins can form active complexes with Cdk2. Cyclin E-Cdk2 kinase is active in the G1 and S phases of the cell cycle and is important (as is Cyclin A-Cdk2) for the progression from G1 to S phase. The levels of Cyclin A-Cdk2 are maximal at the G1/S transition and both Cdk2 and Cyclin A associate with DNA in the initiation complex during replication. The Rb protein has been identified as a substrate for Cdk2-Cyclin E and/or Cdk2-Cyclin A in vivo. This observation is supported by further evidence which shows that Cdk2 is activated and specifically localized to the nucleus during late G1, S phase, and G2.

Spécification

Spécification

Cdk2
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
and ≤0.09% sodium azide., glycerol, Aqueous buffered solution containing BSA
150 μg
Cell Biology
Rat, Mouse, Human, Canine
IgG2a
Western Blot
55
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.