missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A Mouse anti-Human, Unlabeled, Clone: BF683, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 554175

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15877598

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catalytic subunits) to form complexes that regulate the progression of the cell cycle. The main cyclin-cdks complexes formed in vertebrate cells are cyclin D-cdk4 (G0/G1), cyclin E-cdk2 (G1/S), cyclin A-cdk2 (S) and cyclin B1-cdk1 (G2/M). These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small regulatory proteins including p21 and p27Kip1. Specific substrates for cdk-cyclin complexes include nuclear lamins, histones, oncogenes (e.g., c-abl and SV40 large T-Ag), tumor suppressor genes (e.g., retinoblastoma protein, Rb), nucleolin and others. Cyclin A is involved in both S-phase and G2/M transitions of the cell cycle through its association with cdk2 and cdk1, respectively. Cyclin A may also form a complex with the adenovirus oncoprotein E1A which has DNA binding activity. Human cyclin A has been reported to migrate between 54-60kDa by SDS-PAGE and clone BF683 reportedly does not cross-react with mouse, rat or mink cyclin A.

Host Species: Mouse
Clone: BF683
Isotype: IgE
Species Reactivity: Human
Immunogen: Human Cyclin A Recombinant Protein
Formula Weight [Chemical]: 54-60kDa

Flow Cytometry, Immunoprecipitation, Western Blotting

Spécification

Spécification

Cyclin A
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Human Cyclin A Recombinant Protein
0.25 mg
Cell Biology
Human
IgE
Western Blot
BF683
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.