missing translation for 'onlineSavingsMsg'
Learn More

ERK2 Mouse, Unlabeled, Clone: 33, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 610103

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15875259

  • 274.38€ / 1 kit
Date d’expédition estimée: 22-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The family of serine/threonine kinases known as ERKs (extracellular signal regulated kinases) or MAPKs (mitogen-activated protein kinases) are activated after cell stimulation by a wide variety of hormones and growth factors. Cell stimulation induces a signaling cascade that leads to phosphorylation of MEK (MAPK/ERK kinase) which, in turn, activates ERK via tyrosine and threonine phosphorylation. Structural analysis of ERK2 indicates that phosphorylation induces a conformational change that exposes the active site for substrate binding. Myriad proteins represent the downstream effectors for the active ERK and implicate it in the control of cell proliferation and differentiation, as well as regulation of the cytoskeleton. Activation of ERK is normally transient and cells possess dual specificity phosphatases that are responsible for its down-regulation. Furthermore, multiple studies have shown that elevated ERK activity is associated with some cancers. ERK2 is the 42kDa member of the ERK family and is highly homologous to ERK1.

Host Species: Mouse
Clone: 33
Isotype: IgG2b
Species Reactivity: Rat
Immunogen: Rat ERK2 aa. 219-358
Formula Weight [Chemical]: 42kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Spécification

Spécification

ERK2
33
Unconjugated
Affinity Purified
Rat ERK2 aa. 219-358
50μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.