missing translation for 'onlineSavingsMsg'
Learn More

FLK-1 Rat anti-Mouse, APC, Clone: Avas 12alpha1, BD

Rat Monoclonal Antibody

Marque:  BD Biosciences 561993

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15871539

  • 182.58€ / 1 pièce
Date d’expédition estimée: 29-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The Avas 12α1 monoclonal antibody specifically binds to fetal liver kinase 1 (Flk-1), a receptor protein tyrosine kinase closely related to CD117 (c-kit) and CD140a (PDGF Receptor α chain) of the immunoglobulin superfamily. Flk-1, also known as VEGF Receptor-2 (VEGF-R2), is a receptor for vascular endothelial growth factor (VEGF). It is expressed, at the mRNA and protein levels, on distinct sets of mesoderm during gastrulation and on endothelial cells in embryonic and adult tissues. In vivo and in vitro studies indicate that Flk-1 is required for the embryonic development of vascular endothelial and hematopoietic cells.

Spécification

Spécification

FLK-1
Avas 12alpha1
APC
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Fetal liver kinase 1; CD309; Kdr; VEGF receptor-2; VEGFR-2
Affinity Purified
Mouse FLK-1 Recombinant Protein
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.