missing translation for 'onlineSavingsMsg'
Learn More

Foxp3 Rat anti-Mouse, PE, Clone: MF23, BD

Rat Monoclonal Antibody

Marque:  BD Biosciences 560414

Informations supplémentaires : Poids : 0.00930kg

 Afficher plus de versions de ce produit

Code produit. 15870179

  • 190.00€ / 1 pièce
Date d’expédition estimée: 14-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Foxp3 is a 50-55kDa protein also known as Forkhead box P3, JM2, or IPEX. It is a member of the forkhead or winged helix family of transcription factors and is specifically expressed by T regulatory (Treg) cells. Foxp3 has been reported to be a key regulatory protein for Treg cell development and function. Ectopic expression of Foxp3 in conventional T cells is sufficient to induce suppressive activity, repress the production of cytokines such as IL2 and IFN-γ, and upregulate Treg cell-associated molecules such as CD25, CTLA4 and GITR. It has been found that the mutation of Foxp3 is responsible for “scurfy” mice. When overexpressed, Foxp3 leads to poor T cell proliferation and activation.
Monoclonal antibody MF23 reacts with mouse Foxp3. Immunoblotting with MF23 antibody has confirmed it recognizes an epitope between 1-87 amino acids in the N-terminal domain.

Host Species: Rat
Clone: MF23
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse Foxp3 Recombinant Protein

Intracellular Staining

Spécification

Spécification

Foxp3
MF23
PE
Affinity Purified
Mouse Foxp3 Recombinant Protein
25μg
Immunology
Primary
Murine
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.