missing translation for 'onlineSavingsMsg'
Learn More

mGluR5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Thermo Scientific PA138132

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.01000kg

Code produit. 11576313

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

This antibody is predicted to react with bovine, canine, amphibian, mouse, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain tissue.

GRK1 is a member of the guanine nucleotide protein-coupled (G protein) receptor kinase subfamily of Serine/Threonine kinase family. GRK1 phosphorylates rhodopsin and initiates its deactivation. G protein mediated phosphorylation promotes the binding of arrestin, which removes the receptor from the G-protein and terminates the receptor signal chain. GRK1 is farnesylated to promote membrane association. Studies have shown that GRK1 has been shown to be inhibited by recoverin, a Calcium binding protein. Diseases associated with GRK1 protein dysfunction include Oguchi Disease-2 and Hereditary Night Blindness.
Spécification

Spécification

mGluR5
Polyclonal
GRM5
GPRC1E, MGLUR5, mGlu5, metabotropic glutamate receptor 5, metabotropic glutamate receptor 5 variant F, metabotropic glutamate receptor 5 variant G, metabotropic glutamate receptor 5 variant H
Rabbit
Antigen affinity chromatography
RUO
2915, 24418
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
P31424, P41594
GRM5
Synthetic peptide from C-terminus of human mGluR5.
1 mL
Primary
Human, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions