missing translation for 'onlineSavingsMsg'
Learn More

H-2K[d] Mouse anti-Mouse, FITC, Clone: SF1-1.1, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 562003

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15871549

  • 171.36€ / 1 pièce
Date d’expédition estimée: 24-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The SF1-1.1 antibody reacts with the α3 domain of the H-2K[d] MHC class I alloantigen. Reactivity with other haplotypes (e.g, b, j, k, p, q, s, v) has not been observed. It has been reported that plate-bound SF1-1.1 mAb moderately enhances the apoptotic response of thymocytes to plate-bound 145-2C11 mAb (anti-mouse CD3e, Cat. No. 557306/553058).

Spécification

Spécification

H-2K[d]
SF1-1.1
FITC
Affinity Purified
BALB/c mouse cells
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse
IgG2a, κ
RUO
Primary
Murine
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.