Learn More
HLA-A,B,C Mouse anti-Human, FITC, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 555552
Informations supplémentaires : Poids : 0.02000kg
Description
Reacts with a monomorphic epitope of major histocompatibility Class I antigens widely distributed on human nucleated cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G46-2.6
Isotype: IgG1 κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Spécification
HLA-A,B,C | |
Monoclonal | |
FITC | |
Mouse | |
100 Tests | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
G46-2.6 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.