Learn More
IFN-γ Mouse anti-Human, APC, Clone: B27, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 554702
Informations supplémentaires : Poids : 0.00900kg
Description
The B27 antibody reacts with human interferon-γ (IFN-γ). The immunogen used to generate the B27 hybridoma was E.coli-expressed recombinant human IFN-γ. This is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: B27
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human E.coli-expressed IFN-γ
Intracellular Staining
Spécification
IFN-γ | |
Monoclonal | |
0.2mg/mL | |
IFNG; Interferon-gamma; Interferon-γ; Type II interferon; MAF | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
B27 | |
APC | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human E.coli-expressed IFN-γ | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.