missing translation for 'onlineSavingsMsg'
Learn More

Ig κ Light chain Mouse anti-Human, FITC, Clone: G20-193, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 555791

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 15868438

  • 665.00€ / 1 pièce
Expédition estimée: 23-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The G20-193 antibody reacts with human immunoglobulin light chain, kappa (κ). It does not react with λ light chains or heavy chains. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: G20-193
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Spécification

Spécification

Ig κ Light chain
Monoclonal
FITC
Mouse
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
G20-193
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.