Learn More
Ig κ Light chain Mouse anti-Human, PE, Clone: G20-193, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 555792
Informations supplémentaires : Poids : 0.02000kg
Description
The G20-193 antibody reacts with human immunoglobulin light chain, kappa (κ). It does not react with λ light chains or heavy chains. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G20-193
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spécification
Ig κ Light chain | |
Monoclonal | |
PE | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
G20-193 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.