Learn More
Ig κ Light chain Mouse anti-Human, PE, Clone: G20-193, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 562052
Informations supplémentaires : Poids : 0.02000kg
Description
The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa (κ). It does not react with human immunoglobulin μ light chains or human immunoglobulin heavy chains.
Spécification
Ig κ Light Chain | |
Monoclonal | |
PE | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
G20-193 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.