Learn More
IgD Mouse anti-Human, BB515, Clone: IA6-2, BD
Mouse Monoclonal Antibody
218.00€ - 695.00€
Spécification
Antigène | IgD |
---|---|
Clone | IA6-2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | Brilliant Blue 515 (BB515) |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15884179
|
BD Biosciences
565244 |
25 Tests |
218.00€
1 pièce |
Expédition estimée: 15-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15874179
|
BD Biosciences
565243 |
100 Tests |
695.00€
1 pièce |
Expédition estimée: 15-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The IA6-2 monoclonal antibody specifically binds to the heavy chain of human Immunoglobulin D (IgD). IgD is a member of the immunoglobulin superfamily that exists in type 1-membrane (mIgD) and soluble glycoprotein forms. mIgD is expressed on mature naοve B cells (along with membrane IgM) and serves as a B-cell receptor for antigen (BCR). In response to antigen binding, the mIgD BCR, in association with other signaling molecules including CD79a and CD79b, can transduce activating or tolerizing signals intracellularly into B lymphocytes.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Spécification
IgD | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
IgG2a κ | |
Affinity Purified | |
IGHD; Ig delta chain C region; Immunoglobulin heavy constant delta |
IA6-2 | |
Monoclonal | |
Mouse | |
Human | |
Human IgD | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.