missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, κ, Rat, Alexa Fluor™ 488, Clone: R3-34, BD

Marque:  BD Biosciences 557720

Informations supplémentaires : Poids : 0.00840kg

 Afficher plus de versions de ce produit

Code produit. 15879008

  • 452.00€ / 1 pièce
Expédition estimée: 19-04-2024
pour voir le stock disponible 
Ajouter au panier

Description

Description

The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: R3-34
Isotype: IgG1 κ

Flow Cytometry, Intracellular Staining, Isotype Control

Spécification

Spécification

Flow Cytometry, Western Blot
0.2mg/mL
Rat
0.1 mg
IgG1 κ
R3-34
Alexa Fluor 488
Affinity Purified
Undiluted at 4°C
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions