Learn More
anti-IgG1 κ, Rat, FITC, Clone: R3-34, Isotype Control, BD
Marque: BD Biosciences 552916
Informations supplémentaires : Poids : 0.01000kg
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Flow Cytometry, Intracellular Staining, Isotype Control
Spécification
Flow Cytometry, Western Blot | |
FITC | |
Affinity Purified | |
Undiluted at 4°C |
R3-34 | |
Rat | |
50 Tests | |
IgG1 κ |