missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ, Rat, R-PE, Clone: R3-34, Isotype Control, BD
262.00€ - 477.00€
Spécification
Clone | R3-34 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugué | R-PE |
Espèces hôtes | Rat |
Isotype | IgG1 κ |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15857838
|
BD Biosciences
554685 |
0.1 mg |
477.00€
1 pièce |
Expédition estimée: 13-05-2024
Connectez-vous pour voir le stock disponible |
|||||
15866748
|
BD Biosciences
551979 |
50 Tests |
262.00€
1 pièce |
Expédition estimée: 13-05-2024
Connectez-vous pour voir le stock disponible |
|||||
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Flow Cytometry, Intracellular Staining, Isotype Control
Spécification
R3-34 | |
R-PE | |
IgG1 κ | |
Undiluted at 4°C |
Flow Cytometry, Western Blot | |
Rat | |
Affinity Purified |