Learn More
anti-IgG2a κ, Rat , Alexa Fluor™ 647, Clone: R35-95, BD
Marque: BD Biosciences 557906
Informations supplémentaires : Poids : 0.01000kg
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κImmunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Spécification
Flow Cytometry, Western Blot | |
Alexa Fluor 647 | |
Mouse Pooled Immunoglobulin | |
100 Tests | |
IgG2a κ |
R35-95 | |
Rat | |
Affinity Purified | |
Undiluted at 4°C |