Learn More
CD122 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marque: Invitrogen PA547622
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human IL-20 R beta is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD122 Polyclonal antibody specifically detects CD122 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Spécification
CD122 | |
Polyclonal | |
Unconjugated | |
Il2rb | |
CD122; CD122 antigen; high affinity IL-2 receptor beta subunit; high affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL15RB; IL15Rbeta; IL-15Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15Rbeta; IL-2R subunit beta; IL2RB; IL-2RB; IL2RBC; Il-2Rbeta; interleukin 15 receptor, beta; interleukin 2 receptor subunit beta; interleukin 2 receptor, beta; interleukin 2 receptor, beta chain; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70; p70-75; p75 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
16185 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P16297 | |
Il2rb | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-20 R beta Asp14-Asn215 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |