Learn More
IL-10 Rat anti-Human, PE, Clone: JES3-19F1, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 559330
Informations supplémentaires : Poids : 0.02000kg
Description
The JES3-19F1 antibody reacts with human (IL-10). The immunogen used to generate the JES3-19F1 hybridoma was recombinant human IL-10 expressed in COS cells. This is a neutralizing antibody. This antibody also reacts with viral IL-10. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Intracellular Staining
Spécification
IL-10 | |
Monoclonal | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
JES3-19F1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Recombinant Human IL-10 | |
100 Tests | |
Immunology | |
Human, Virus | |
IgG2a |
For Research Use Only.