missing translation for 'onlineSavingsMsg'
Learn More

IL-17A Mouse anti-Human, Alexa Fluor 488, Clone: N49-653, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 560488

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 15870239

  • 482.00€ / 1 pièce
Date d’expédition estimée: 14-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Human IL-17A, also known as IL-17, is a proinflammatory cytokine that is encoded by the IL17A gene in chromosome 6. IL-17A is produced as a disulfide-linked homodimer comprised of two mature 136-amino acid polypeptides. It is a member of the IL-17 family of structurally related cytokines, designated IL-17A through IL-17F. Activated memory T cells, especially Th17 cells (specialized IL-17A-producing CD4+ T cells distinct from Th1 and Th2 cells) produce IL-17 and provide protective immunity against pathogens. Activated CD8+ T cells, γδT cells, NK cells and neutrophils can also be activated to produce IL-17A. IL-17A binds to and exerts its biological activity through IL-17 receptors (IL-17R) that are expressed by a variety of target cells including fibroblasts, epithelial and endothelial cells, monocytes/macrophages and mast cells. The ubiquitous IL-17R expression pattern may explain the broad tissue responsiveness to IL-17. IL-17 induces stromal cells to secrete cytokines and chemokines involved in inflammatory and hematopoietic processes. For example, IL-17 induces fibroblasts to produce IL-6, IL-8, G-CSF and express increased surface ICAM-1. The N49-653 antibody reacts with human IL-17A.

Host Species: Mouse
Clone: N49-653
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IL-17A Recombinant Protein

Intracellular Staining

Spécification

Spécification

IL-17A
N49-653
IL-17; CTLA8; Cytotoxic T-lymphocyte-associated serine esterase 8
Affinity Purified
Human IL-17A Recombinant Protein
100 Tests
Immunology
Primary
Human
Flow Cytometry
Alexa Fluor 488
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.