missing translation for 'onlineSavingsMsg'
Learn More

IL-4 Mouse anti-Human, Unlabeled, Clone: 8D4-8, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 556917

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15868798

  • 259.00€ / 1 pièce
Date d’expédition estimée: 17-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody reacts with human interleukin-4 (IL-4). The immunogen used to raise the 8D4-8 hybridoma was recombinant human IL-4. The 8D4-8 antibody binds to an epitope that is different than the epitope recognized by the MP4-25D2 antibody (Mfr. No.554485).

Host Species: Mouse
Clone: 8D4-8
Isotype: IgG1, κ
Cross Reactivity: Human

Blocking Assay, ELISA, Intracellular Staining, Western Blotting

Spécification

Spécification

IL-4
8D4-8
Unconjugated
Affinity Purified
Recombinant Human IL-4
0.1mg
Immunology
Primary
Human
ELISA
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.