Learn More
IL-4 Rat anti-Mouse, PE, Clone: 11B11, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 562044
Informations supplémentaires : Poids : 0.01000kg
Description
The 11B11 monoclonal antibody specifically binds to mouse Interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 prepared from the supernatant of Phorbol 12-Myristate 13-Acetate (PMA)-stimulated EL-4 cells. The 11B11 antibody is reportedly a neutralizing antibody.
Spécification
IL-4 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Partially Purified Mouse IL-4 | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
11B11 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 |
For Research Use Only.