missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Receptor Monoclonal Antibody (B-R6), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen AHR0061
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.50000kg
Description
IL-6 Receptor Monoclonal antibody specifically detects IL-6 Receptor in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization.Spécification
IL-6 Receptor | |
Monoclonal | |
4.0 mg/mL | |
PBS with no preservative; pH 7.2 | |
P08887 | |
IL6R | |
CHO cells transfected with cDNA corresponding to the gp80 IL-6 receptor gene. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization | |
B-R6 | |
Unconjugated | |
IL6R | |
CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
Mouse | |
purified | |
RUO | |
3570 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |