missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL10RA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA535361
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
IL10RA Polyclonal antibody specifically detects IL10RA in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Spécification
IL10RA | |
Polyclonal | |
Unconjugated | |
Il10ra | |
AW553859; CD210; CD210a; CDw210; CDW210A; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; HIL-10R; I10R; I10R1; il-10 receptor; il-10 receptor antagonist; IL-10 receptor subunit alpha; IL10R; IL-10R alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10R1; IL-10R2; Il10ra; IL-10RA; interleukin 10 receptor subunit alpha; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha; mIL-10R | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3587 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q13651 | |
Il10ra | |
KLH conjugated synthetic peptide between 147-175 amino acids from the central region of human IL10RA | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |