missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B NP Monoclonal Antibody (B017 (B35G)), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA180712
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Mouse anti Influenza B Nucleoprotein antibody, clone B017 recognizes an epitope within the nucleoprotein of influenza B virus.
Influenza B NP Monoclonal antibody specifically detects Influenza B NP in Virus samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spécification
Influenza B NP | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide; pH 7.5 | |
Mouse | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry, Western Blot | |
B017 (B35G) | |
Unconjugated | |
Influenza B; Influenza NP | |
Influenza B/Lee/40 and B/Singapore/-222/79 viruses. | |
1 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |