Learn More
Ly-6G and Ly-6C Rat anti-Mouse, PerCP-Cy5.5, Clone: RB6-8C5, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 552093
Informations supplémentaires : Poids : 0.01000kg
Description
The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation. The antigen is also expressed on the monocyte lineage in the bone marrow, but not on erythroid cells. In the periphery, RB6-8C5 antibody recognizes granulocytes (neutrophils and eosinophils) and monocytes. The RB6-8C5 mAb is a component of the “lineage cocktail” used in studies of hematopoietic lineages. The mAb 1A8 (Cat. No. 551461) specifically recognizes Ly-6G, but not Ly-6C.
Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b κ
Species Reactivity: Mouse
Flow Cytometry
Spécification
Ly-6G and Ly-6C | |
Monoclonal | |
0.2mg/mL | |
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
RB6-8C5 | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Not Reported | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
For Research Use Only.