missing translation for 'onlineSavingsMsg'
Learn More

MAP1B Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 612679

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15876259

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Microtubule-associated proteins (MAPs) play a crucial role in the development and structure of nerve cells. These proteins are important for the assembly and stability of microtubles during neurite outgrowth and for the morphology of neuronal processes, such as dendrites. MAP1B encodes for a single precursor protein that is cleaved to produce a single heavy chain of 280kDa and a light chain of 32kDa which become noncovalently associated. Expression levels of MAP1B are reportedly highest in the brain and are modulated during development as reported with gene silencing experiments that implicate MAP1B's role during neuronal differentiation. In addition, MAP1B can be selectively phosphorylated depending on the cell type, subcellular localization and developmental phase. In neurons, MAP1B is expressed as a glycoprotein at the plasma membrane, where it can interact with proteins participating in axonal guiding. MAP1B has been reported to be observable to migrate in a range between 320-340kDa.

Western Blotting

Spécification

Spécification

MAP1B
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse MAP1B aa. 1745-1858
150 μg
Cell Biology
Human, Mouse, Rat
IgG2a
Western Blot
6
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.