missing translation for 'onlineSavingsMsg'
Learn More

MEK2 Mouse, Unlabeled, Clone: 96/MEK2, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 610236

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15875349

  • 497.76€ / 1 kit
Date d’expédition estimée: 22-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

MEK (Map/Erk Kinase) 1 and 2 are serine/threonine kinases, also known as MAP kinase kinases (MAP2K1 and 2, MAPKK1 and 2, or MKK1 and 2). They activate the MAP (ctivated Protein) kinases, also known as ERKs (Extracellular signal Regulated Kinases), which are critical kinases in multiple signal transduction pathways that regulate cell growth and differentiation. Activation of MEK 1 and 2 is dependent upon phosphorylation of serines 218 and/or 222 by activated MAP kinase kinase kinases (MAP3Ks), such as the Raf isoforms. Hormones, growth and differentiating factors, or tumor promoters induce Raf activation via activation of Ras proteins. MEK2 is seven amino acids larger and shares 81% identity with MEK1. In cultured cells, MEK2 is activated by serum. In vitro, v-Raf phosphorylates and activatesMEK2. It is thought that all of these activated protein kinases are downstream of the Ras signal transduction pathway and represent an integral part of the Ras mitogenic signal.

The 96/MEK2 monoclonal antibody recognizes MEK2, regardless of phosphorylation status.

Spécification

Spécification

MEK2
96/MEK2
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
MAP Kinase Kinase 2; ERK Kinase
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.