missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA533822
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
mGluR4 Polyclonal antibody specifically detects mGluR4 in Bovine, Equine, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spécification
mGluR4 | |
Polyclonal | |
Unconjugated | |
GRM4 | |
G protein-coupled receptor GRM4; G protein-coupled receptor, family C, group 1, member D; glutamate metabotropic receptor 4; glutamate receptor; glutamate receptor, metabotropic 4; GPRC1D; GRM4; Metabotropic glutamate receptor 4; mGlu4; MGLUR4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100061879, 100152616, 100347907, 24417, 268934, 2914, 790333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P31423, Q14833, Q68EF4 | |
GRM4 | |
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GRM4 / MGLUR4. | |
50 μg | |
Primary | |
Bovine, Equine, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
Antibody | |
IgG |