missing translation for 'onlineSavingsMsg'
Learn More

MICAL1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen PA530804

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.01000kg

Code produit. 13485307

  • 474.00€ / 100µL
Expédition estimée: 22-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Recommended positive controls: HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MICAL1 Polyclonal antibody specifically detects MICAL1 in Human samples. It is validated for Western Blot.
Spécification

Spécification

MICAL1
Polyclonal
Unconjugated
MICAL1
[F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1
Rabbit
Antigen affinity chromatography
RUO
64780
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q8TDZ2
MICAL1
Recombinant protein fragment corresponding to a region within amino acids 175 and 391 of Human MICAL1
100 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions