missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIF Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA518383
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
MIF Polyclonal antibody specifically detects MIF in Human samples. It is validated for Immunohistochemistry, Western Blot.Spécification
MIF | |
Polyclonal | |
Unconjugated | |
Mif | |
cytokine; delayed early response protein 6; DER6; GIF; GLIF; glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; HGNC:7097; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migration inhibitory factor; macrophage migration inhibitory factor (glycosylation-inhibiting factor); MGC107654; Mif; MMIF; phenylpyruvate tautomerase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4282 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P14174 | |
Mif | |
Synthetic peptide sequence (NAANVGWNNSTFA) corresponding to the C-terminus amino acids of MIF | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |