Learn More
MINA53 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA511157
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
MINA protein is directly involved in ribosome biogenesis, most likely during the assembly process of preribosomal particles. This protein is also involved in cell proliferation. MINA may have a role in esophageal squamous cell carcinoma, colon cancer and lung cancer.Spécification
MINA53 | |
Polyclonal | |
RIOX2 | |
Protein mina53; MYC-induced nuclear antigen; Mineral dust-induced gene protein 2; MINA; MDIG; mina53; | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
84864 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8IUF8 | |
RIOX2 | |
KLH conjugated synthetic peptide between 397-429 amino acids from the C-terminal region of human MINA | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |