Learn More
MINT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA1072
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
PA1-072 detects munc-18 interacting protein 3 (Mint3) from mouse cells and rat brain samples. PA1-072 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽86 kDa protein representing Mint3 from AtT20 cell extract and rat brain samples. The PA1-072 immunogen is a synthetic peptide corresponding to residues M(1) E F L P E P Q H P P G P P T M D L E(19) of rat Mint3. The PA1-072 immunizing peptide (Cat. # PEP-135) is available for use in neutralization and control experiments.
MINT3 Polyclonal antibody specifically detects MINT3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spécification
MINT3 | |
Polyclonal | |
Unconjugated | |
APBA3 | |
Adapter protein X11gamma; amyloid beta (A4) precursor protein-binding family A member 3 (X11-like 2); amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2); amyloid beta (A4) precursor protein-binding, family A, member 3; amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2); amyloid beta A4 precursor protein-binding family A member 3; amyloid beta precursor protein binding family A member 3; Amyloid-beta A4 precursor protein-binding family A member 3; Apba3; AW208791; lin 10; lin-10; MGC:15815; Mint 3; MINT3; mint-3; Neuronal Munc18-1-interacting protein 3; neuron-specific X11L2 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; X11gamma; X11L2; X11-like 2 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57267, 83611, 9546 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O70248, O88888, O96018 | |
APBA3 | |
Synthetic peptide corresponding to residues M(1) E F L P E P Q H P P G P P T M D L E(19) of rat Mint3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |