Learn More
MITD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA543864
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Peptide sequence: RAENIKKYLD QEKEDGKYHK QIKIEENATG FSYESLFREY LNETVTEVWI Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 82%
MITD1 Polyclonal antibody specifically detects MITD1 in Human samples. It is validated for Western Blot.Spécification
MITD1 | |
Polyclonal | |
Unconjugated | |
MITD1 | |
1500032H18Rik; hypothetical protein MGC56159; microtubule interacting and trafficking domain containing 1; MIT domain-containing protein 1; MIT, microtubule interacting and transport, domain containing 1; Mitd1; RGD1307700; zgc:56159 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
129531 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WV92 | |
MITD1 | |
Synthetic peptide directed towards the middle region of human MITD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |