missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MiTF Monoclonal Antibody (21D1418), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA516214
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.10000kg
Description
Suggested positive control: A375 or RAW.
MiTF Monoclonal antibody specifically detects MiTF in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spécification
MiTF | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
O75030, O88368, Q08874 | |
MITF | |
A synthetic peptide mapping to the 300-350 amino acid region of MITF. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
21D1418 | |
Unconjugated | |
MITF | |
BCC2; Bhlhe32; black eyed white; bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanocyte inducing transcription factor; melanogenesis associated transcription factor; mi; microphtalmia-associated transcription factor; microphthalmia; microphthalmia associated transcription factor isoform M-; microphthalmia transcription factor; microphthalmia-associated transcription factor; microphthalmia-associated transcription factor A; microphthalmia-associated transcription factor isoform H; microphthalmia-associated transcription factor isoform H protein; microphthalmia-associated transcription factor Trnc-1; microphthalmia-associated transcription factor Trnc-2; microphthalmia-associated transcription factor variant 1; microphthalmia-associated transcription factor variant 2; microphthalmia-associated transcription factor variant 3; microphthalmia-associated transcription factor variant MITF-M(+); MITF; MITF-H; MITF-M; MITF-M-; transcription factor, isoform 7; vit; Vitiligo; Wh | |
Mouse | |
Protein G | |
RUO | |
17342, 25094, 4286 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |