Learn More
PARP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA14280
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
PA1-4280 detects the PARP-2 protein in human samples. PA1-4280 has been successfully used in Western blot procedures. This antibody detects a ∽66 kDa protein representing PARP-2 in 3T3 cell lysate. PA1-4280 immunizing peptide corresponds to amino acid residues 43-59 of the 62 kDa human PARP-2 protein. Store at 4oC for 3 months or at -20oC for longer periods.
PARP2 Polyclonal antibody specifically detects PARP2 in Human samples. It is validated for Western Blot.Spécification
PARP2 | |
Polyclonal | |
Unconjugated | |
PARP2 | |
ADP-ribosyltransferase (NAD+, poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 2; ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADPRT2; ADPRT-2; ADPRTL2; ADPRTL3; ARTD2; Aspartl2; C78626; DNA ADP-ribosyltransferase PARP2; HGNC:272; hPARP-2; mPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; parp 2; PARP2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribosyl) transferase-like 2; poly [ADP-ribose] polymerase 2; poly(ADP-ribose) polymerase 2; poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum with 0.02% sodium azide | |
Q9UGN5 | |
PARP2 | |
Synthetic peptide corresponding to residues Q(43) R Q E S K K M P V A G G K A N K(59) of human PARP2. | |
RUO | |
10038 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |