Learn More
ADCY10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA543049
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Peptide sequence: VGHTVRHEYT VIGQKVNLAA RMMMYYPGIV TCDSVTYNGS NLPAYFFKEL Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
ADCY10 Polyclonal antibody specifically detects ADCY10 in Human samples. It is validated for Western Blot.Spécification
ADCY10 | |
Polyclonal | |
Unconjugated | |
ADCY10 | |
3',5'-cyclic AMP synthetase; 4930431D04Rik; 4931412F17; Adcy10; adenylate cyclase 10; adenylate cyclase 10 (soluble); adenylate cyclase 10, soluble; Adenylate cyclase homolog; Adenylate cyclase type 10; AH-related protein; ATP pyrophosphate-lyase; epididymis secretory sperm binding protein Li 7a; germ cell soluble adenylyl cyclase; HCA2; HEL-S-7a; hsAC; RP1-313L4.2; Sac; SACI; Sacy; soluble adenylyl cyclase; testicular soluble adenylyl cyclase; testicular soluble adenylyl cyclase (SAC) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55811 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PN6 | |
ADCY10 | |
Synthetic peptide directed towards the N-terminal of human SAC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |