missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI/SR-BII Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA316805
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
SR-BI/SR-BII Polyclonal antibody specifically detects SR-BI/SR-BII in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot.Spécification
SR-BI/SR-BII | |
Polyclonal | |
Unconjugated | |
SCARB2 | |
85 kDa lysosomal membrane sialoglycoprotein; 85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2; 9330185J12Rik; AI120173; AMRF; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2 (lysosomal integral membrane protein II); CD36 antigen-like 1; CD36 antigen-like 2; CD36L1; CD36L2; Cla1; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; EPM4; HDL QTL 1; Hdlq1; HDLQTL6; Hlb398; HLGP85; LGP85; LIMP II; LIMP2; LIMP-2; LIMPII; lysosome membrane protein 2; Lysosome membrane protein II; MLGP85; mSR-BI; Scarb1; Scarb2; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; Scavenger receptor class B member | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot | |
Conc. Not Determined | |
whole serum with 0.02% sodium azide | |
O35114, P27615, P97943, Q14108, Q61009, Q8WTV0 | |
SCARB1, SCARB2 | |
Extracellular residues 230-380 of Scavenger Receptor-BI/BII expressed as two tandem copies in bacteria using the pET system. | |
RUO | |
117106, 12492, 20778, 25073, 949, 950 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |