Learn More
PSF Progressive Safety™ Anti-static Safety Wellington Boots To EN345
Marque: PSF Progressive Safety™ AGRIP/W-4
Informations supplémentaires : Poids : 1.00000kg
Description
- Antislip
- Anti-static
- Waterproof rubber sole
- Resistant to heat, oils, acids & animal fats
- 200 joule toe protection
- White
Spécification
4 |