Learn More
αβ T-Cell Receptor Mouse anti-Rat, FITC, Clone: R73, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 561672
Informations supplémentaires : Poids : 0.02000kg
Description
The R73 antibody reacts with the αβ T-cell Receptor (TCR) found on most peripheral T lymphocytes, intestinal intraepithelial lymphocytes, and thymocytes. It does not react with γδ TCR-bearing cells. Cross-linked R73 mAb induces T-cell differentiation and activation. In vivo treatment with mAb R73 can suppress immune function of peripheral αβ TCR-expressing T cells, and reduce the severity of experimental autoimmune, transplant rejection, and graft-versus-host responses.
Spécification
αβ T-Cell Receptor | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat T blasts and rat erythrocytes | |
50 μg | |
Immunology | |
Rat | |
IgG1 κ |
Flow Cytometry | |
R73 | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.