missing translation for 'onlineSavingsMsg'
Learn More

TLS Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 611385

Informations supplémentaires : Poids : 0.01100kg

 Afficher plus de versions de ce produit

Code produit. 15865909

  • 605.34€ / 1 kit
Expédition estimée: 20-02-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Chromosomal translocations in leukemias and malignancies of non-hematopoietic tissues usually result in the fusion of genes and the production of novel fusion proteins. Translocated in liposarcoma (TLS/FUS) was identified through its fusion to the transcription factor CHOP in human myxoid liposarcoma. In addition, the fusion of TLS to the transcription factor ERG is involved in human acute myeloid leukemia. TLS contains an N-terminal Gln-, Ser-, and Tyr-rich region (QSY) that is thought to be a potent transactivator when fused with transcription factors. In its C-terminus, TLS contains a ribonucleoprotein consensus sequence (RNP-CS) and Arg-Gly-Gly (RGG) repeats that have been implicated in RNA binding. This feature allows it to function in shuttling of RNA from the nucleus. TLS also interacts with Ser-Arg proteins that regulate RNA splicing. Interestingly, TLS also interacts with the DNA binding domains of the estrogen, thyroid hormone, and glucocorticoid receptors. Thus, TLS is a multifunctional protein that is involved in RNA transport, nuclear receptor function, and gene transactivation when fused with transcription factors.

Immunofluorescence, Western Blotting

Spécification

Spécification

TLS
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human TLS aa. 2-117
150 μg
Cell Biology
Rat, Human
IgG1
Western Blot
15
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.