Learn More
TNF Mouse anti-Human, PE-Cy7, Clone: MAb11, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 560923
Informations supplémentaires : Poids : 0.01000kg
Description
The MAb11 monoclonal antibody specifically binds to human tumor necrosis factor (TNF, also known as TNF-α) protein. TNF is an efficient juxtacrine, paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The MAb11 antibody has been reported to crossreact with Rhesus Macaque TNF.
Spécification
TNF | |
Monoclonal | |
PE-Cyanine7 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Recombinant Human TNF | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MAb11 | |
Tumor necrosis factor alpha; TNF-a; TNF-α; TNFSF2; Cachectin | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.