missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM1L1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA518896
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
TOM1L1 Polyclonal antibody specifically detects TOM1L1 in Human samples. It is validated for Western Blot.Spécification
TOM1L1 | |
Polyclonal | |
Unconjugated | |
TOM1L1 | |
2310045L10Rik; C80573; Jerry protein; OK/KNS-CL.3; RGD1562626; Src activating and signaling molecule; src-activating and signaling molecule protein; SRCASM; target of myb1 (chicken)-like 1; target of myb1 like 1 membrane trafficking protein; target of myb1-like 1; target of myb1-like 1 (chicken); Target of Myb-like protein 1; TOM1L1; TOM1-like protein 1; Unknown (protein for MGC:142395) | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10040 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75674 | |
TOM1L1 | |
Synthetic peptide sequence (EQNKNQKEATNTTSE) corresponding to the internal amino acids of TOM1L1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |